Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Torso and head

Deluxe torso and head in 28 parts, 8 kg, 870×380×250 mm
  763-0096
Marketsource
Torso and head
  763-0096
Marketsource
Torso and head
Torso and head

Deluxe torso and head in 28 parts, 8 kg, 870×380×250 mm

 
967,00 €
   
1 * 1 ST
 

Very detailed torso for advanced demonstrations.

  • The following parts are removable and divisible: spine, chest, head, heart, lungs, stomach, liver, kidneys and more

Interchangeable genitalia, open back and neck section show vertebrae, discs, nerves, blood vessels and more, 4-part brain.

 
  • Weight:
    8 kg
  • Description:
    Deluxe torso and head in 28 parts
  • W×D×H:
    870×380×250 mm