Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flexible skeleton model

Flexible skeleton model

Skeleton with movable spine, height: 1700 mm, 8 kg
Catalog # 763-0074
Marketsource
Supplier:   3B Scientific
Flexible skeleton model
Catalog # 763-0074
Supplier:  3B Scientific
Marketsource
Flexible skeleton model
Flexible skeleton model

Skeleton with movable spine, height: 1700 mm, 8 kg

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Skeleton of unbreakable plastic with joint ligaments on the right skeleton half. The ligaments faithfully reproduced are found in the shoulder, elbow, knee and ankle joints (763-0072) or the same skeleton with a movable and flexible spine. Almost all natural movements can be demonstrated. Visible spinal nerves, blood vessels and intervertebral discs (discs) (763-0074).

  • Skeleton with ligament or with movable spine

Height 170 cm, weight 8 kg.

Specifications
  • Weight:
    8 kg
  • Description:
    Skeleton with movable spine, height: 1700 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.