Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample Bag, A5/C5, COEX

Sample Bag, A5/C5, COEX

Sample bags, Black, Plastic, 165×245, 70 µm
Catalog # DAKL9057
Supplier:   DAKLAPACK
Sample Bag, A5/C5, COEX
Catalog # DAKL9057
Supplier:  DAKLAPACK
Sample Bag, A5/C5, COEX

Sample bags, Black, Plastic, 165×245, 70 µm

Price
41,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Coex bag A5/C5 165×245 with a 50 mm flap is ideal for protecting various products such as clothing, small equipment and miscellaneous items during the shipment.

  • Super strong, 70 micron thick, extra thick quality
  • Equipped with a permanent closing strip
  • The coex bag is made of opaque light gray plastic with a black inside
  • The coex bag can also be used as a shipping envelope, shipping by letterbox post is possible (note the maximum thickness)
Specifications
  • Colour:
    Black
  • Description:
    Sample bags
  • L×W:
    165×245
  • Material:
    Plastic
  • Thickness:
    70 µm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.