ContactUsLinkComponent
Research® plus 4-Pack with epT.I.P.S.® BioBased
Research® plus 4-Pack with epT.I.P.S.® BioBased
Catalog # EPPE3123000.950
Supplier:  EPPENDORF
undefined
Research® plus 4-Pack with epT.I.P.S.® BioBased
Catalog # EPPE3123000.950
Supplier:  EPPENDORF
Specifications
  • Description:
    Eppendorf Research® plus, 4-pack set with BioBased epT.I.P.S.® pipette tips
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This convenient set offers safe, ergonomic and lightweight pipettes along with an abundance of high-quality filter tips made from bio-based material (except filter). Package includes 4 Eppendorf Research® plus pipettes and 3840 matching ep Dualfilter T.I.P.S.® BioBased tips, covering a volume range of 0,1 to 1000 μl.
  • ACT now bundle
  • Eppendorf Research® plus pipettes are renowned for their high reliability as well as lightweight and ergonomic design, making them the standard equipment in thousands of laboratories worldwide
  • Set comprises of 4 ACT-labeLled single-channel volume variants and ample stocks of sterile filter tips
  • Ideal for laboratories engaged in sensitive micro- or molecular biological applications
  • Included in this package are high-quality, ep Dualfilter T.I.P.S.® BioBased filter pipette tips, fully made from an innovative bio-based polymer (except filter) significantly reducing the product-related carbon footprint, thereby contributing to a more sustainable laboratory environment without compromising experimental results
  • Certified free from human DNA, DNase, RNase, and PCR inhibitors, tips ensure optimal performance in PCR applications
Delivery: Includes 4 pipettes, 3840 pipette tips, 4 pipette tip boxes and a ballpoint pen:
Eppendorf™ Research™ plus 1-channel dark gray pipette with variable volume 0,1 to 2,5 μl
Eppendorf™ Research™ plus 1-channel yellow pipette with variable volume 2 to 20 μl
Eppendorf™ Research™ plus 1-channel yellow pipette with variable volume 20 to 200 μl
Eppendorf™ Research™ plus 1-channel blue pipette with variable volume 100 to 1000 μl
ep Dualfilter T.I.P.S.™ BioBased Reloads, PCR clean/Sterile, 0,1 to 10 μl M, 40 mm, medium gray, colourless tips, 960 tips (10 trays×96 tips)
ep Dualfilter T.I.P.S.™ BioBased Reloads, PCR clean/Sterile, 2 to 20 μl, 53 mm, yellow, colourless tips, 960 tips (10 trays×96 tips)
ep Dualfilter T.I.P.S.™ BioBased Reloads, PCR clean/Sterile, 2 to 200 μl, 55 mm, yellow, colourless tips, 960 tips (10 trays×96 tips)
ep Dualfilter T.I.P.S.™ BioBased Reloads, PCR clean/Sterile, 50 to 1000 μl, 76 mm, blue, colourless tips, 960 tips (10 trays×96 tips)
3× epT.I.P.S.™ Box 2.0 (empty box for 10 to 300 μl)
1× epT.I.P.S.™ Box 2.0 (empty box for 1000 to 1250 μl)
1× Eppendorf™ pipette pen