Orders LinkContactUsLinkComponent
Evaluation Software for Medical Application
Evaluation Software for Medical Application
Winlog medical software for routine checks, Pharmaceutical and medical application
Catalog # EBRO1340-2460
Supplier:   EBRO
Evaluation Software for Medical Application
Catalog # EBRO1340-2460
Supplier:  EBRO
Evaluation Software for Medical Application
Winlog medical software for routine checks, Pharmaceutical and medical application
Price
1 470,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For routine checks.
  • User friendly
  • High-precision measurements
  • Automatic report generation
  • Automatic user-defined calculations
  • Automatic recognition of process cycles
  • Current version: 3.71
Supports: EBI 10, EBI 12, EBI 11, EBI 16, EBI 20, EBI 40, EBI 100, EBI 300, EBI 310. Creation of user-defined templates for specific devices and thermal processes. Three-dimensional display of sensor placement. FDA 21 CFR Part 11 functionality also possible.
Hardware requirements: Processor speed minimum 1,6 GHz, Working memory 2 GB (32 bit), Working memory 4 GB (64 bit) 4 GB free hard disc space, USB A (universal serial bus).
Software requirements: Windows vista 8 (32 bit or 64 bit), windows 10 (32 bit or 64 bit).
Specifications
  • Description:
    Winlog medical software for routine checks
  • For:
    Pharmaceutical and medical application