ContactUsLinkComponent
Haematoxylin, crystals for microscopy (C.I. 75290), Sigma-Aldrich®
Irritant
Haematoxylin, crystals for microscopy (C.I. 75290), Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Haematoxylin, crystals for microscopy (C.I. 75290), Sigma-Aldrich®
Supplier:  Merck
Specification Test Results
  • Identity (UV/VIS-Spectrum):
    passes test
  • Transition range:
    pH 5.0 - 7.2, yellow - violet
  • Water (according to Karl Fischer):
    ≤ 10 %
  • Suitability for microscopy:
    passes test
  • Papanicolaous stain (cyanophilic Cytoplasm):
    blue-green to green
  • Papanicolaous stain (eosinophilic Cytoplasm):
    pink
  • Papanicolaous stain (Erythrocytes):
    red
  • Papanicolaous stain (Cell nuclei):
    blue to dark violet
  • Papanicolaous stain (Microorganisms):
    grey-blue
  • H&E staining (Cell nuclei):
    dark blue to dark violet
  • H&E staining (Cytoplasma):
    pink to red
  • H&E staining (intercellular substances):
    pink to red
  • H&E staining (Erythrocytes):
    yellow to orange