Order Entry
Belgium
Orders LinkContactUsLinkComponent
trans-4-Hydroxy-L(-)-proline

trans-4-Hydroxy-L(-)-proline

Glass bottle for solids, 250 g
Catalog # A1705.0250
Supplier:   PanReac AppliChem
trans-4-Hydroxy-L(-)-proline
Catalog # A1705.0250
Supplier:  PanReac AppliChem
CAS Number:  51-35-4
trans-4-Hydroxy-L(-)-proline

Glass bottle for solids, 250 g

CAS Number:  51-35-4
Price
837,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  • CAS Number:51-35-4
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064320
  • Molecular Weight:131.13 g/mol
  • Merck Index:13,04866
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:206 °C
  • Storage Temperature:Ambient
  • EINECS:200-091-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.