(R)-(-)-Mandelic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:611-71-2
:
Formula:C₈H₈O₃
MDL Number:MFCD00064251
:152.15 g/mol
Boiling Pt:322 °C (1013 hPa)
Melting Pt:130…132 °C
Density:1.341 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>190 °C
EINECS:210-276-6
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:130 - 132 °C
- Molar Mass:152.15 g/mol
- Bulk Density:670 kg/m3
- Flash Point:190 °C
- pH:2.1 (60 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together