Order Entry
Belgium
ContactUsLinkComponent
Haematoxylin monohydrate, Certistain® for microscopy, Sigma-Aldrich®
Haematoxylin monohydrate, Certistain® for microscopy, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Haematoxylin monohydrate, Certistain® for microscopy, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Hill Formula:
    C₁₆H₁₄O₆ * H₂O
  • Molar Mass:
    320.29 g/mol
  • Melting Point:
    140 °C Elimination of water of crystallisation
  • Bulk density:
    500 kg/m³
  • Solubility:
    (20 °C) slightly soluble
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 2 water endangering

Specification Test Results

Related Information

Frequently Bought Together