Orders LinkContactUsLinkComponent
Conductivity Sensor, Smartec CLD18
Conductivity Sensor, Smartec CLD18
Conductivity sensor, Smartec CLD18, stainless steel housing, IP 69, NEMA6P (body), Tri-clamp ISO 2852 50,8 mm
Catalog # ENDRCLD18-1014/0
Supplier:   Endress+Hauser
Conductivity Sensor, Smartec CLD18
Catalog # ENDRCLD18-1014/0
Supplier:  Endress+Hauser
Conductivity Sensor, Smartec CLD18
Conductivity Sensor, Smartec CLD18
Conductivity sensor, Smartec CLD18, stainless steel housing, IP 69, NEMA6P (body), Tri-clamp ISO 2852 50,8 mm
Price
2 470,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Compact conductivity system for hygienic applications.
  • The compact design allows installation in narrow pipes without obstructing the flow
  • Effortlessly integrates into your plant infrastructure with the optional IO-Link protocol
  • The product's unique hygienic design prevents contamination, has 3-A approval and complies with EG regulations 2023/2006 and 1935/2004
  • Designed for cleaning-in-place (CIP)
 : For optional items or more information, contact your Avantor representative.
 : CLD18 compact conductivity system.
Specifications
  • Connection:
    Tri-clamp ISO 2852 50,8 mm
  • Description:
    Conductivity sensor, Smartec CLD18, stainless steel housing, IP 69, NEMA6P (body)