Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

DL-Proline

Glass bottle for solids, 5 g
Catalog # MOLEM67043584
Marketsource
Supplier:   Molekula
DL-Proline
Catalog # MOLEM67043584
Supplier:  Molekula
Marketsource
CAS Number:  609-36-9
DL-Proline

Glass bottle for solids, 5 g

CAS Number:  609-36-9
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (±)-Proline, H-DL-Pro-OH
  • CAS Number:609-36-9
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00005250
  • Molecular Weight:115.13 g/mol
  • Merck Index:12,07963
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:208…210 °C
  • Density:1.38 g/cm³ (22.9 °C)
  • EINECS:210-189-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications