DL-Proline 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (±)-Proline, H-DL-Pro-OH
- CAS Number:609-36-9
- Formula:C₅H₉NO₂
- MDL Number:MFCD00005250
- Molecular Weight:115.13 g/mol
- Merck Index:12,07963
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:208…210 °C
- Density:1,38 g/cm³ (22.9 °C)
- EINECS:210-189-3
2 Options Available Below
Product Details & Documents
Synonyms: (±)-Proline, H-DL-Pro-OH
- CAS Number:609-36-9
- Formula:C₅H₉NO₂
- MDL Number:MFCD00005250
- Molecular Weight:115.13 g/mol
- Merck Index:12,07963
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:208…210 °C
- Density:1,38 g/cm³ (22.9 °C)
- EINECS:210-189-3
Documents
ACRO17403
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Sulfated ash:≤0.1 %
- Solubility:(5 % in water) Colorless to light yellow
- Titration with HClO4:98.5 to 101.5 %
- Heavy metals (as Pb):≤20 ppm



