Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Lecithin

Marketsource
Supplier:   Roth Carl
Lecithin
Supplier:  Roth Carl
Marketsource
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:8002-43-5
  • MDL Number:MFCD00147406
  • Merck Index:12,05452
  • Density:1.0305 g/cm³ (20 °C)
  • EINECS:232-307-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 20,50 €
    20,50 €
    1 * 100 g
       20,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 37,50 €
    37,50 €
    1 * 250 g
       37,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 112,50 €
    112,50 €
    1 * 1 kg
       112,50 €

    Specifications:
    Marketsource
    Marketsource