Order Entry
Belgium
ContactUsLinkComponent
Lecithin ∼90% (from soy bean)
Lecithin ∼90% (from soy bean)
Catalog # A2218.0100
Supplier:  PanReac AppliChem
CAS Number:  
Lecithin ∼90% (from soy bean)
Catalog # A2218.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
EINECS:310-129-7
CAS Number:8030-76-0
MDL Number:MFCD00147406