FMOC-Lys(5-FITC)-OH for biomolecular labeling
Catalog # AATB5070
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
:757.81 g/mol
ADR:5,II
:GHS07
CAS Number:1095493-05-2