Skip to Header
Skip to Main Content
Skip to Footer
Avancé
Avancé
Fiches de données de sécurité (FDS)
Certificats
Données du fabricant
0
0,00 €
Order Entry
Login
/
S'inscrire
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy5.5 biotin conjugate
Cy5.5 biotin conjugate biodetection
Cy5.5 biotin conjugate biodetection
Irritant
Cy5.5 biotin conjugate biodetection
Catalog # AATB3105
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Cy5.5 biotin conjugate biodetection
Catalog # AATB3105
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
1299.45 g/mol
ADR:
5,II
:
GHS07
Product Details