Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-Biotin-N-succinimidyl ester
D-Biotin-N-succinimidyl ester, ReadiView™ biodetection
D-Biotin-N-succinimidyl ester, ReadiView™ biodetection
D-Biotin-N-succinimidyl ester, ReadiView™ biodetection
AATB3059
:
AAT BIOQUEST
:
D-Biotin-N-succinimidyl ester, ReadiView™ biodetection
AATB3059
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
341.38 g/mol
:
5,II
:
GHS07
:
35013-72-0
:
C₁₄H₁₉N₃O₅S
:
MFCD00078531