Order Entry
Belgium
ContactUsLinkComponent
 
Cyanine 3 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy3® bisNHS ester]
Catalog # AATB270
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 3 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy3® bisNHS ester]
Catalog # AATB270
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :949.1 g/mol
CAS Number:1311966-47-8
Formula:C43H49KN4O14S2