
Tide Quencher™ 1 phosphoramidite, Tide Quencher™ fluorescent probe
Catalog # AATB2198
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 µmol
- Seq:9140
Specifications
About this item
:
:589.73 g/mol
ADR:5,II
:GHS07