
DiSBAC2(3) for membrane potential measurement
Catalog # AATB21414
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:436.55 g/mol
ADR:5,II
:GHS07
CAS Number:47623-98-3
Formula:C₁₉H₂₄N₄O₄S₂