Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
Lucigenin chloride indicator
Catalog # AATB21259
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Lucigenin chloride indicator
Catalog # AATB21259
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :510.51 g/mol
ADR:5,II
 :GHS07
CAS Number:2315-97-1
 :22103-92-0
Formula:C₂₈H₂₂N₄O₆
MDL Number:MFCD00011925