0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Tide Quencher™ 6WS amine
Tide Quencher™ 6WS amine, Tide Quencher™ fluorescent probe
Tide Quencher™ 6WS amine, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 6WS amine, Tide Quencher™ fluorescent probe
Catalog # AATB2091
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Tide Quencher™ 6WS amine, Tide Quencher™ fluorescent probe
Catalog # AATB2091
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
981.11 g/mol
ADR:
5,II
:
GHS07
Product Details