Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TF4-dUTP
TF4-dUTP 1 mM in TRIS buffer (pH 7.5)
TF4-dUTP 1 mM in TRIS buffer (pH 7.5)
AATB17009
:
AAT Bioquest
:
TF4-dUTP 1 mM in TRIS buffer (pH 7.5)
AATB17009
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
25 Nmo
Seq:
0311
:
:
1250 g/mol
:
5,II
:
GHS07