Order Entry
Belgium
ContactUsLinkComponent
Cyanine 5 bissuccinimidyl ester ≥90% [equivalent to Cy5® bisNHS ester]
Cyanine 5 bissuccinimidyl ester ≥90% [equivalent to Cy5® bisNHS ester]
Catalog # AATB157
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 5 bissuccinimidyl ester ≥90% [equivalent to Cy5® bisNHS ester]
Catalog # AATB157
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1038.23 g/mol