Cyanine 3 alkyne ≥95% [equivalent to Cy3® alkyne]
:
:781.86 g/mol
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Bottles, PET, Square, with Screw Cap
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
: VWR Collection
Microplates, 96-well with glass inserts
: HIRSCHMANN
Volumetric measuring scoops, SteriWare®
: Sampling Systems
ZEONOR WIDTH 200 UM 100 µM LID THICKNESS 1 * 1 Pce
: MARKET SOURCE PART PROCESS
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
: EPPENDORF
VWR®, Cryo Boxes
: VWR Collection
Cell culture inserts for multiwell plates, ThinCert™
: Greiner Bio-One