Order Entry
Belgium
ContactUsLinkComponent
TAMRA-cGMP PDE V substrate "Red Fluorescence"
Irritant
TAMRA-cGMP PDE V substrate "Red Fluorescence"
Catalog # AATB13605
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
TAMRA-cGMP PDE V substrate "Red Fluorescence"
Catalog # AATB13605
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    0,5 µmol
  • Seq:
    9030

Specifications

About this item

 :
ADR:5,II
 :GHS07