ContactUsLinkComponent
ReadiUse™ Spin Filter (5), 10KD
  AATB60502
undefined
ReadiUse™ Spin Filter (5), 10KD
  AATB60502
  • Description:
    ReadiUse™ 10KD spin filter
  • Pk:
    5
ReadiUse™ 10 Kd spin filter is a disposable ultrafiltration device for the concentration of biological samples such as antibodies.
  • Compatible with both swing bucket or fixed angle instruments
  • Designed to prevent membrane fouling
  • Low internal surface area
  • High material recovery from dilute solutions
 : For research use only.