Order Entry
Belgium
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 546)
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 546)
Catalog # AATB16537
Supplier:  AAT Bioquest
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 546)
Catalog # AATB16537
Supplier:  AAT Bioquest

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Clone:
    IgG
  • Conjugation:
    iFluor® 546
  • Host:
    Goat
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Mouse
  • Cross adsorption:
    Min × Human
  • Pk:
    200 µG

Specifications

About this item

iFluor® 546 is a bright orange fluorescent dye.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins
Caution: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse