HIS Lite™ Recombinant Bis NTA-Ni Complex, Cy5
Catalog # AATB12613
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Type de conditionnement:Vial
- Cdt:1 mg
- Conjugaison:Cy5®
- Type de protéine/peptide:Recombinant
- Synonyme de protéine:Nitrilotriacetic Acid
- Nom de la protéine/du peptide:Bis NTA-Ni
- Molecular weight:1500,3
Specifications
About this item
The Cy5 Bis NTA-Ni Complex can be directly applied either to an SDS-PAGE gel or western blot membrane for fluorescence imaging.
- Specific and highly-sensitive detection
- Minimal crossreactivity
No secondary reaction is required since the Ni-NTA complexes are directly conjugated to the fluorophores.
Caution: For research use only.