Order Entry
Belgium
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), PerCP (Peridinin-Chlorophyll Protein Complex)-iFluor™ 647
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), PerCP (Peridinin-Chlorophyll Protein Complex)-iFluor™ 647
Catalog # AATB16906
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), PerCP (Peridinin-Chlorophyll Protein Complex)-iFluor™ 647
Catalog # AATB16906
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 µG
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-iFluor™ 647
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin, Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

Specifications

About this item

Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.

  • Extremely high biotin-binding affinity
  • High quality conjugate
Caution: For research use only.