Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 514
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
- Optimized for flow cytometry
- Useful for assay development
- High affinity
- Multiple applications
: For research use only.
- Conjugation:iFluor® 514
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin
- Protein/peptide name:Streptavidin
Frequently Bought Together