Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Violet 540
AATB16932
:
- Pack type:Vial
- Pk:100 µG
- Conjugation:mFluor™ Violet 540
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin
- Protein/peptide name:Streptavidin
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
- Optimized for flow cytometry
- Useful for assay development
- High affinity
- Multiple applications
: For research use only.