Order Entry
Belgium
ContactUsLinkComponent
Native Amanita phalloides Phalloidin (from Eukaryotic cells), TMR (Tetramethylrhodamine)
Native Amanita phalloides Phalloidin (from Eukaryotic cells), TMR (Tetramethylrhodamine)
Catalog # AATB23102
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Amanita phalloides Phalloidin (from Eukaryotic cells), TMR (Tetramethylrhodamine)
Catalog # AATB23102
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    300 Tests
  • Conjugation:
    TMR (Tetramethylrhodamine)
  • Protein/peptide type:
    Native
  • Source:
    Eukaryotic cells
  • Species:
    Amanita phalloides
  • Protein synonyms:
    Amanita phalloides
  • Protein/peptide name:
    Phalloidin

Specifications

About this item

This orange fluorescent phalloidin conjugate (equivalent to TRITC-labeled phalloidin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.

  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.