Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nuclear Red™ DCS1

Nuclear Red™ DCS1

Nuclear Red™ DCS1, 0,5 mL
N° de catalogue AATB17552
Fournisseur:   AAT Bioquest
Nuclear Red™ DCS1
N° de catalogue AATB17552
Fournisseur:  AAT Bioquest
Numéro CAS:  
Nuclear Red™ DCS1
Nuclear Red™ DCS1

Nuclear Red™ DCS1, 0,5 mL

Numéro CAS:  
Prix
156,00 €
Log in for availability
1 * 0,5 mL
Product Details & Documents

Our Nuclear Red™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analysing DNA content in dead, fixed or apoptotic cells.

  • Cell-impermeable
  • Useful for multicolour imaging
  • Bright fluorescence

5 mM DMSO solution.

 : For research use only.

Specifications

Specifications

  • Pk:
    0,5 mL
  • Description:
    Nuclear Red™ DCS1
  • Cat. No.:
    AATB17552
  • Purity:
    95%

Specifications