ReadiUse™ Microscope Mounting Solution (50 ml)
ReadiUse™ microscope mounting solution, 50 mL
Catalog # AATB20009
Supplier: AAT Bioquest
ReadiUse™ Microscope Mounting Solution (50 ml)
Catalog # AATB20009
Supplier: AAT Bioquest
CAS Number:
Jump to:
Product Details & Documents
The mounting medium is the solution in which the specimen is embedded, generally under a cover glass.
- Non-reactive
- Long-term stability
- Minimal hands-on time
- Ready to use
Caution: For research use only.
Specifications
Specifications
- Pk:50 mL
- Description:ReadiUse™ microscope mounting solution



