Saisie de commande
Belgium
ContactUsLinkComponent
ReadiUse™ Cell Detaching Buffer (50 ml)
ReadiUse™ Cell Detaching Buffer (50 ml)
Catalog # AATB60010
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiUse™ Cell Detaching Buffer (50 ml)
Catalog # AATB60010
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Cdt:
    50 mL
  • Désignation:
    ReadiUse™ cell detaching buffer

Specifications

About this item

ReadiUse™ cell detaching buffer is a cell detachment solution that does not contain mammalian or bacterial-derived products.

  • Compatible with all major instruments
  • No mammalian or bacterial products
  • Low toxicity
  • High effectiveness
Caution: For research use only.