TET-dT Phosphoramidite
Supplier: AAT BIOQUEST
Certificates
About this item
:
Molecular Weight:1563.35 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together
Accessories for Q Cyclers
Supplier: Quantabio
Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles
Supplier: Thermo Fisher Scientific
Sampling spoons with long handle, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
U96 DeepWell™ plates 1,3 and 2,0 ml, Nunc®
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Reagent/centrifuge tubes, round bottom, with or without push cap
Supplier: SARSTEDT
Closures, PPCO, natural, with colour coder, 11 mm
Supplier: Thermo Fisher Scientific
Ice pans, Bel-Art Magic Touch 2™
Supplier: Bel-Art Products, a Part of SP