Order Entry
Belgium
ContactUsLinkComponent
ReadiLink™ Rapid iFluor® 700 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
ReadiLink™ Rapid iFluor® 700 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1245
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ Rapid iFluor® 700 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1245
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid iFluor™ 700 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'

Specifications

About this item

AAT Bioquest's iFluor® dyes are developed for labelling proteins, in particular, antibodies.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.