ContactUsLinkComponent
Amplite® Colorimetric Urea Quantitation Kit *Blue Color*
Amplite® Colorimetric Urea Quantitation Kit *Blue Color*
Catalog # AATB10058
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Colorimetric Urea Quantitation Kit *Blue Color*
Catalog # AATB10058
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric urea quantitation kit *blue color*
  • Tests per kit:
    200 Tests
Specifications
About this item
Urea is the final degradation product of protein and amino acid metabolism in animals.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.