ContactUsLinkComponent
Amplite® Fluorimetric Total Thiol Quantitation Assay Kit *Green Fluorescence*
Amplite® Fluorimetric Total Thiol Quantitation Assay Kit *Green Fluorescence*
Catalog # AATB5524
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Total Thiol Quantitation Assay Kit *Green Fluorescence*
Catalog # AATB5524
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ fluorimetric total thiol quantitation assay kit *green fluorescence*
  • Tests per kit:
    200 Tests
Specifications
About this item
The detection and measurement of free thiol (such as free cysteine, glutathione and cysteine residues in proteins) is one of the essential tasks for investigating biological processes and events in many biological systems.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.