ContactUsLinkComponent
Helixyte™ Green Fluorometric dsDNA Quantitation kit *Optimised For Microplate Readers*
Helixyte™ Green Fluorometric dsDNA Quantitation kit *Optimised For Microplate Readers*
Catalog # AATB17650
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Helixyte™ Green Fluorometric dsDNA Quantitation kit *Optimised For Microplate Readers*
Catalog # AATB17650
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Helixyte™ green fluorimetric dsDNA quantitation kit *optimized for microplate readers*
Specifications
About this item
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.