Order Entry
Belgium
ContactUsLinkComponent
PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Blue Fluorescence
PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Blue Fluorescence
Catalog # AATB21611
Supplier:  AAT Bioquest
CAS Number:  
undefined
PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Blue Fluorescence
Catalog # AATB21611
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    PhosphoWorks™ fluorimetric pyrophosphate assay kit *blue fluorescence*

Specifications

About this item

Pyrophosphate (PPi) is produced by a number of biochemical reactions, such as ATP hydrolysis, DNA and RNA polymerizations, cyclic AMP formation by the enzyme adenylate cyclase and the enzymatic activation of fatty acids to form their coenzyme A esters.

  • Pyrophosphate-dependent fluorescence
  • Highly selective for PPi
  • Contains all necessary components
Caution: For research use only.