ContactUsLinkComponent
Cell Navigator® Mitochondrion Staining Kit *Red Fluorescence*
Cell Navigator® Mitochondrion Staining Kit *Red Fluorescence*
Catalog # AATB22668
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Navigator® Mitochondrion Staining Kit *Red Fluorescence*
Catalog # AATB22668
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    500 Assays
  • Description:
    Cell Navigator™ mitochondrion staining kit *red fluorescence*
Specifications
About this item
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labelling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.