Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*
Catalog # AATB22971
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Microplate Reader*
Catalog # AATB22971
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ fluorimetric mitochondrial superoxide activity assay kit *optimized for microplate reader*

Specifications

About this item

Mitochondria are major producers of cellular superoxide.

  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimised for microplate readers but suitable for all major fluorescence platforms
  • results with only 1 hour incubation
Caution: For research use only.