Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Flow Cytometry*
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22804
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22804
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ mitochondrion membrane potential assay kit *orange fluorescence optimized for flow cytometry*

Specifications

About this item

This particular kit is designed to detect cell apoptosis by measuring the loss of the mitochondrial membrane potential(MMP).

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.