ContactUsLinkComponent
Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers
Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers
Catalog # AATB11111
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers
Catalog # AATB11111
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Portelite™ fluorimetric protein quantitation Kit *Optimized for CytoCite™ and Qubit™ Fluorometers*
Specifications
About this item
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
  • Dynamic range from 12.5 ug/ml to 5 mg/ml of bsa
  • Optimised for cytocite™ and qubit™ fluorometers
  • Highly sensitive
  • Robust protocol
Caution: For research use only.