Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Fluorimetric Cellular Voltage Assay Kit
Cell Meter™ Fluorimetric Cellular Voltage Assay Kit
Catalog # AATB35000
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fluorimetric Cellular Voltage Assay Kit
Catalog # AATB35000
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ fluorimetric cellular voltage assay kit

Specifications

About this item

Almost all plasma membranes have an electrical potential across them, with the inside usually negative with respect to the outside.

  • Contains all necessary components
  • Multiple research applications
  • Ratiometric detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.