ContactUsLinkComponent
Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*
Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*
Catalog # AATB20130
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*
Catalog # AATB20130
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Cell Meter™ live cell caspase 3/7 Imaging kit *green fluorescence*
Specifications
About this item
Cell Meter™ Live Cell No Wash Caspase 3/7 Imaging Kit uses our recently developed cell-permeable fluorogenic caspase substrate, ApoSight™ Green Caspase 3/7, the first fluorogenic probe, for the direct detection of caspase activities in live cells.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.