Order Entry
Belgium
ContactUsLinkComponent
ReadiLink™ Rapid XFD647 Antibody Labelling Kit *XFD647 Same Structure to Alexa Fluor™ 647* (2×50 µg Labellings)
ReadiLink™ Rapid XFD647 Antibody Labelling Kit *XFD647 Same Structure to Alexa Fluor™ 647* (2×50 µg Labellings)
Catalog # AATB1278
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ Rapid XFD647 Antibody Labelling Kit *XFD647 Same Structure to Alexa Fluor™ 647* (2×50 µg Labellings)
Catalog # AATB1278
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Description:
    ReadiLink™ rapid XFD647 antibody labelling kit 'XFD647 same structure to Alexa Fluor™ 647'

Specifications

About this item

XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.