Cell Meter™ Cellular Senescence Activity Assay Kit *Green Fluorescence*
Catalog # AATB23005
Supplier: AAT Bioquest
CAS Number:
Specifications
- Cdt:100 Tests
- Désignation:Cell Meter™ cellular senescence activity assay kit *green fluorescence*
Specifications
About this item
Cellular Senescence is an irreversible growth arrest triggered in order to prevent growth in DNA damaged cells.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.